(±)-Cbz-α-phosphonoglycine trimethyl ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-Benzyloxycarbonyl-2-phosphonoglycine trimethyl ester, (±)-Benzyloxycarbonyl-α-phosphonoglycine trimethyl ester, (±)-Methyl 2-benzyloxycarbonylamino-2-(dimethoxyphosphinyl)acetate, Methyl 2-{[(benzyloxy)carbonyl]amino}-2-(dimethoxyphosphoryl)acetate, Methyl 2-(((benzyloxy)carbonyl)amino)-2-(dimethoxyphosphoryl)acetate
B22506-06
B22506-14
CAAAB22506-03EA
164.56
CAD
CAAAB22506-03
CAAAB22506-06
CAAAB22506-14
(±)-Cbz-α-phosphonoglycine trimethyl ester 97%
(±)-Cbz-α-phosphonoglycine trimethyl ester
Purity: 97%
CAS No.: 88568-95-0
Molecular Formula: C13H18NO7P
Formula Weight: 331.27
Melting Point: 76-80°
MDL No.: MFCD00043304
Formula:
C₁₃H₁₈NO₇P MW: 331.27 g/mol Melting Pt: 76…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043304 CAS Number: 88568-95-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...