4-Cyano-4'-pentyloxybiphenyl 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Amyloxy-4'-cyanobiphenyl, 4-Cyano-4'-n-pentyloxybiphenyl, 5OCB, 4'-Amyloxy-4-biphenylcarbonitrile, 4'-Pentyloxy-4-biphenylcarbonitrile, 4'-Pentyloxy-[1,1'-biphenyl]-4-carbonitrile
B22477-03
B22477-06
CAAAB22477-03EA
100.65
CAD
CAAAB22477-03
CAAAB22477-06
4-Cyano-4'-pentyloxybiphenyl 99%
4-Cyano-4'-pentyloxybiphenyl
Purity: 99%
CAS No.: 52364-71-3
Molecular Formula: C18H19NO
Formula Weight: 281.42
Flash Point:
Formula:
C₁₈H₁₉NO MW: 265.35 g/mol Melting Pt: 48 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00074879 CAS Number: 52364-71-3 EINECS: 257-875-9 UN: 3439 ADR: "6.1",III |
Learn more
![](/m-ca.vwr.com/en_CA/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...