N-Fmoc-L-norleucine 98%

Supplier: Thermo Scientific Chemicals
Warning

B22475-03 B22475-06 B22475-14
CAAAB22475-03EA 67.62 CAD
CAAAB22475-03 CAAAB22475-06 CAAAB22475-14
N-Fmoc-L-norleucine 98%
N-Fmoc-L-norleucine

Purity: 98% CAS No.: 77284-32-3 Molecular Formula: C21H23NO4 Formula Weight: 353.42 Storage Temperature: 2°C to 8°C Melting Point: 140-142° MDL No.: MFCD00037537

Formula: C₂₁H₂₃NO₄
Melting Pt: 140…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00037537
CAS Number: 77284-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR