2-(1H-Imidazol-5-yl)acetic acid hydrochloride 97%
Supplier: Thermo Scientific Chemicals
B22473-03
B22473-06
CAAAB22473-03EA
173.8
CAD
CAAAB22473-03
CAAAB22473-06
2-(1H-Imidazol-5-yl)acetic acid hydrochloride 97%
2-(1H-Imidazol-5-yl)acetic acid hydrochloride
A GABAc antagonist
Purity: 97%
CAS No.: 3251-69-2
Molecular Formula: C5H6N2O2.HCl
Formula Weight: 162.58
Odor: Odorless
Melting Point: 221-224°
Beilstein Reference: 3701591
MDL No.: MFCD00012698
Formula:
C₅H₇ClN₂O₂ Melting Pt: 221…224 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012698 CAS Number: 3251-69-2 EINECS: 221-840-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...