Bicyclo[3.2.0]hept-2-en-6-one 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(±)-cis-Bicyclo[3.2.0]hept-2-en-6-one
B22472-06
B22472-22
CAAAB22472-06EA
178.54
CAD
CAAAB22472-06
CAAAB22472-14
CAAAB22472-22
Bicyclo[3.2.0]hept-2-en-6-one 97%
Bicyclo[3.2.0]hept-2-en-6-one
Purity: 97%
CAS No.: 13173-09-6
Molecular Formula: C7H8O
Formula Weight: 108.14
UN No.: 1224
MDL No.: MFCD00066201
Formula:
C₇H₈O MW: 108.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00066201 CAS Number: 13173-09-6 UN: 1224 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...