4-Cyanophenyl-4-hexylbenzoate 99%

Supplier: Thermo Scientific Chemicals
Warning

B22471-03 B22471-06
CAAAB22471-03EA 49.45 CAD
CAAAB22471-03 CAAAB22471-06
4-Cyanophenyl-4-hexylbenzoate 99%
4-Cyanophenyl 4-hexylbenzoate

Purity: 99% CAS No.: 50793-85-6 Molecular Formula: C20H21NO2 Formula Weight: 307.39 Melting Point: 44-47° MDL No.: MFCD00600506

Formula: C₂₀H₂₁NO₂
Melting Pt: 44…47 °C
Storage Temperature: Ambient
MDL Number: MFCD00600506
CAS Number: 50793-85-6
EINECS: 256-767-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR