6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

B22469-06 B22469-14 B22469-22
CAAAB22469-06EA 63.52 CAD
CAAAB22469-06 CAAAB22469-14 CAAAB22469-22
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride

Purity: 97% CAS No.: 2328-12-3 Molecular Formula: C11H15NO2.HCl Formula Weight: 229.71 Melting Point: 256-260° Beilstein Reference: 3634126

Formula: C₁₁H₁₆ClNO₂
MW: 229.71 g/mol
Melting Pt: 256…260 °C
Storage Temperature: Ambient
MDL Number: MFCD00012744
CAS Number: 2328-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR