Ethyl thiophene-3-carboxylate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl 3-Thiophenecarboxylate, 3-Thiophenecarboxylic acid ethyl ester

B22468-03 B22468-06 B22468-14
CAAAB22468-03EA 59.57 CAD
CAAAB22468-03 CAAAB22468-06 CAAAB22468-14
Ethyl thiophene-3-carboxylate 98%
Ethyl thiophene-3-carboxylate

Purity: 98% CAS No.: 5751-80-4 Molecular Formula: C7H8O2S Formula Weight: 156.20 Refractive Index: 1.5225 Boiling Point: 207-208° MDL No.: MFCD02179226

Formula: C₇H₈O₂S
MW: 156.20 g/mol
Boiling Pt: 207…208 °C
Storage Temperature: Ambient
MDL Number: MFCD02179226
CAS Number: 5751-80-4
EINECS: 227-275-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR