4-(Methylthio)benzyl alcohol 98%
Supplier: Thermo Scientific Chemicals
B22455-06
B22455-14
B22455-22
CAAAB22455-06EA
80.26
CAD
CAAAB22455-06
CAAAB22455-14
CAAAB22455-22
4-(Methylthio)benzyl alcohol 98%
4-(Methylthio)benzyl alcohol
Purity: 98%
CAS No.: 3446-90-0
Molecular Formula: C8H10OS
Formula Weight: 154.23
Flash Point:
Formula:
C₈H₁₀OS Boiling Pt: 108…109 °C/0.4 mm Melting Pt: 41…43 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009706 CAS Number: 3446-90-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...