O,O'-Di-2-pyridyl thiocarbonate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22451-03
B22451-06
B22451-14
CAAAB22451-03EA
131.3
CAD
CAAAB22451-03
CAAAB22451-06
CAAAB22451-14
O,O'-Di-2-pyridyl thiocarbonate 98%
O,O'-Di-2-pyridyl thiocarbonate
Purity: 98%
CAS No.: 96989-50-3
Molecular Formula: C11H8N2O2S
Formula Weight: 232.26
Storage Temperature: -30°C to -10°C
Melting Point: 96-99°
MDL No.: MFCD00074870
Formula:
C₁₁H₈N₂O₂S Melting Pt: 96…99 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00074870 CAS Number: 96989-50-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...