2-Hydroxy-4-(methacryloyloxy)benzophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

B22445-06 B22445-14 B22445-22
CAAAB22445-06EA 148.68 CAD
CAAAB22445-06 CAAAB22445-14 CAAAB22445-22
2-Hydroxy-4-(methacryloyloxy)benzophenone 99%
2-Hydroxy-4-(methacryloyloxy)benzophenone

Purity: 99% CAS No.: 2035-72-5 Molecular Formula: C17H17O4 Formula Weight: 282.30 Storage Temperature: 2°C to 8°C Melting Point: 77-79° MDL No.: MFCD00080561

Formula: C₁₇H₁₄O₄
MW: 282.30 g/mol
Melting Pt: 77…79 °C
Density: 1.18
Storage Temperature: Ambient
MDL Number: MFCD00080561
CAS Number: 2035-72-5
EINECS: 218-000-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR