2-(Trifluoromethyl)acrylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B22443-03 B22443-06 B22443-14
CAAAB22443-03EA 102.07 CAD
CAAAB22443-03 CAAAB22443-06 CAAAB22443-14
2-(Trifluoromethyl)acrylic acid 98%
2-(Trifluoromethyl)acrylic acid

Purity: 98% CAS No.: 381-98-6 Molecular Formula: C4H3F3O2 Formula Weight: 140.06 Flash Point: 82°(180°F) Melting Point: 49-51° UN No.: 3261 MDL No.: MFCD00042424

Formula: C₄H₃F₃O₂
Melting Pt: 49…51 °C
Flash Pt: 82 °C(180 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042424
CAS Number: 381-98-6
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR