2-(tert-Butoxycarbonylthio)-4,6-dimethylpyrimidine 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Boc-thio-4,6-dimethylpyrimidine, O-tert-Butyl S-(4,6-dimethylpyrimidin-2-yl) carbonothioate

B22435-06 B22435-14
CAAAB22435-06EA 35.71 CAD
CAAAB22435-06 CAAAB22435-14
2-(tert-Butoxycarbonylthio)-4,6-dimethylpyrimidine 97%
2-(tert-Butoxycarbonylthio)-4,6-dimethylpyrimidine

Purity: 97% CAS No.: 41840-28-2 Molecular Formula: C11H16N2O2S Formula Weight: 240.33 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 48-51° Beilstein Reference: 396984 MDL No.: MFCD00006080

Formula: C₁₁H₁₆N₂O₂S
MW: 240.32 g/mol
Melting Pt: 48…51 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006080
CAS Number: 41840-28-2
EINECS: 255-562-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR