Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22429-06
B22429-14
B22429-22
CAAAB22429-06EA
38.39
CAD
CAAAB22429-06
CAAAB22429-14
CAAAB22429-22
Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate 98%
Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate
Purity: 98%
CAS No.: 5909-24-0
Molecular Formula: C8H9ClN2O2S
Formula Weight: 232.69
Storage Temperature: -30°C to -10°C
Melting Point: 60-63°
MDL No.: MFCD00006085
Formula:
C₈H₉ClN₂O₂S Melting Pt: 60…63 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006085 CAS Number: 5909-24-0 EINECS: 227-619-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...