1,3,5-Triethynylbenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

B22417-03 B22417-06
CAAAB22417-03EA 279.66 CAD
CAAAB22417-03 CAAAB22417-06
1,3,5-Triethynylbenzene 98%
1,3,5-Triethynylbenzene

Purity: 98% CAS No.: 7567-63-7 Molecular Formula: C12H6 Formula Weight: 150.18 Melting Point: 105-107° UN No.: 1325 MDL No.: MFCD00068621

Formula: C₁₂H₆
Melting Pt: 105…107 °C
Density: 1.05
Storage Temperature: Ambient
MDL Number: MFCD00068621
CAS Number: 7567-63-7
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR