2,3,3-Trimethylindolenine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22410-06
B22410-14
CAAAAB22410-06EA
59.09
CAD
CAAAAB22410-06
CAAAAB22410-14
2,3,3-Trimethylindolenine 98%
2,3,3-Trimethylindolenine
Purity: 98%
CAS No.: 1640-39-7
Molecular Formula: C11H13N
Formula Weight: 159.23
Flash Point: 93°(199°F)
Refractive Index: 1.5470
Melting Point: 6-8°
Boiling Point: 228-229°
Beilstein Reference: 119740
MDL No.: MFCD00005724
Formula:
C₁₁H₁₃N MW: 159.23 g/mol Boiling Pt: 228…229 °C Melting Pt: 6…8 °C Density: 0,992 Flash Pt: 93 °C (199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005724 CAS Number: 1640-39-7 EINECS: 216-685-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...