N-(tert-Butoxycarbonyl)-4-aminobutyric acid ≥98%

Supplier: Thermo Scientific Chemicals

B22407-03 B22407-06
CAAAB22407-03EA 54.04 CAD
CAAAB22407-03 CAAAB22407-06
N-(tert-Butoxycarbonyl)-4-aminobutyric acid ≥98%
N-(tert-Butoxycarbonyl)-4-aminobutyric acid

Purity: 98+% CAS No.: 57294-38-9 Molecular Formula: (CH3)3COCONH(CH2)3CO2H Formula Weight: 203.24 Melting Point: 57-60° MDL No.: MFCD00037313

Formula: C₉H₁₇NO₄
Melting Pt: 57…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00037313
CAS Number: 57294-38-9
EINECS: 628-652-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR