Ethylene glycol monohexyl ether 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22385-03
B22385-06
CAAAB22385-03EA
89.9
CAD
CAAAB22385-03
CAAAB22385-06
Ethylene glycol monohexyl ether 99%
Ethylene glycol monohexyl ether
Purity: 99%
CAS No.: 112-25-4
Molecular Formula: C8H18O2
Formula Weight: 146.23
Flash Point: 94°(201°F)
Refractive Index: 1.4330
Melting Point: -42°
Boiling Point: 208°
Beilstein Reference: 1734691
UN No.: 2922
MDL No.: MFCD00045997
Formula:
C₈H₁₈O₂ Boiling Pt: 208 °C Melting Pt: –42 °C Density: 0.888 Flash Pt: 94 °C (201 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00045997 CAS Number: 112-25-4 EINECS: 203-951-1 UN: 2922 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...