N-Benzyloxycarbonyl-D-alaninol 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(2R)-N-Benzyloxycarbonylamino-1-propanol, N-Cbz-D-Alaninol, Z-D-Alaninol, (R)-2-(Cbz-amino)-1-propanol
B22372-03
B22372-06
B22372-14
CAAAB22372-03EA
34.6
CAD
CAAAB22372-03
CAAAB22372-06
CAAAB22372-14
N-Benzyloxycarbonyl-D-alaninol 98%
N-Cbz-D-Alaninol
Purity: 98%
CAS No.: 61425-27-2
Molecular Formula: C11H15NO3
Formula Weight: 209.25
Storage Temperature: 2°C to 8°C
Melting Point: 82-84°
MDL No.: MFCD00672531
Formula:
C₁₁H₁₅NO₃ MW: 209.2 g/mol Melting Pt: 82…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00672531 CAS Number: 61425-27-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...