Methallyl phenyl ether 96%
Supplier: Thermo Scientific Chemicals
B22369-06
B22369-14
CAAAB22369-06EA
78.05
CAD
CAAAB22369-06
CAAAB22369-14
Methallyl phenyl ether 96%
Methallyl phenyl ether
Purity: 96%
CAS No.: 5820-22-4
Molecular Formula: C10H12O
Formula Weight: 148.21
Flash Point: 77°(171°F)
Refractive Index: 1.5150
Boiling Point: 204°
Beilstein Reference: 1933868
MDL No.: MFCD00008597
Formula:
C₁₀H₁₂O |
MDL Number:
MFCD00008597 CAS Number: 5820-22-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...