2-(Methylthio)naphthalene 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Methyl(naphthalen-2-yl)sulfane, Methyl(naphthalen-2-yl)sulphane
B22368-03
B22368-06
CAAAB22368-03EA
180.12
CAD
CAAAB22368-03
CAAAB22368-06
CAAAB22368-14
2-(Methylthio)naphthalene 98%
2-(Methylthio)naphthalene
Purity: 98%
CAS No.: 7433-79-6
Molecular Formula: C11H10S
Formula Weight: 174.27
Odor: Stench
Melting Point: 62-63°
Beilstein Reference: 1859409
MDL No.: MFCD00051615
Formula:
C₁₁H₁₀S MW: 174.27 g/mol Melting Pt: 62…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051615 CAS Number: 7433-79-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...