Ethyl-5-bromothiophene-2-carboxylate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 5-bromothiophene-2-carboxylate
B22355-03
B22355-06
CAAAB22355-03EA
73.79
CAD
CAAAB22355-03
CAAAB22355-06
CAAAB22355-14
Ethyl-5-bromothiophene-2-carboxylate 99%
Ethyl 5-bromothiophene-2-carboxylate
Purity: 99%
CAS No.: 5751-83-7
Molecular Formula: C7H7BrO2S
Formula Weight: 235.10
Boiling Point: 94-96°/4mm
MDL No.: MFCD02683089
Formula:
C₇H₇BrO₂S MW: 235.10 g/mol Boiling Pt: 94…96 °C/4 mm Storage Temperature: Ambient |
MDL Number:
MFCD02683089 CAS Number: 5751-83-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...