2-Cyclopentylethanol 98+%
Supplier: Thermo Scientific Chemicals
Warning
|
B22344-03
B22344-06
CAAAB22344-03EA
139.2
CAD
CAAAB22344-03
CAAAB22344-06
2-Cyclopentylethanol 98+%
2-Cyclopentylethanol
Purity: 98+%
CAS No.: 766-00-7
Molecular Formula: C7H14O
Formula Weight: 114.19
Flash Point: 70°(158°F)
Refractive Index: 1.4585
Boiling Point: 182°
Beilstein Reference: 2069860
MDL No.: MFCD00019301
Formula:
C₇H₁₄O Boiling Pt: 182 °C Density: 0.920 Flash Pt: 70 °C(158 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00019301 CAS Number: 766-00-7 EINECS: 212-156-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...