Azo rubine

Supplier: Thermo Scientific Chemicals

Synonyms: Carmoisine B, acid Red 14, Brilliant Crimson Red, Sodium 4-hydroxy-3-((4-sulfonatonaphthalen-1-yl)diazenyl)naphthalene-1-sulfonate, Sodium 4-hydroxy-3-((4-sulphonatonaphthalen-1-yl)diazenyl)naphthalene-1-sulphonate

B22328-09 B22328-18 B22328-30
CAAAB22328-09EA 71.1 CAD
CAAAB22328-09 CAAAB22328-18 CAAAB22328-30
Azo rubine
Azo rubine

CAS No.: 3567-69-9 Molecular Formula: C20H12N2Na2O7S2 Formula Weight: 502.42 MDL No.: MFCD00003978

Formula: C₂₀H₁₄N₂O₇S₂
Storage Temperature: Ambient
MDL Number: MFCD00003978
CAS Number: 3567-69-9
EINECS: 222-657-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR