L-Aspartic acid monosodium salt monohydrate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium (S)-3-amino-3-carboxypropanoate hydrate, Sodium L-aspartate monohydrate, L-Aspartic acid sodium salt monohydrate

B22321-22 B22321-36
CAAAB22321-22EA 83.74 CAD
CAAAB22321-22 CAAAB22321-36
L-Aspartic acid monosodium salt monohydrate 99%
L-Aspartic acid monosodium salt monohydrate

Purity: 99% CAS No.: 323194-76-9 Molecular Formula: C4H6NNaO4.H2O Formula Weight: 173.11 (155.10anhy) MDL No.: MFCD00152960

Formula: C₄H₆NNaO₄·H₂O
MW: 173.11 g/mol (155.10 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00152960
CAS Number: 323194-76-9
EINECS: 223-264-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR