1-(p-Toluenesulfonyl)indole 95%
Supplier: Thermo Scientific Chemicals
B22320-03
B22320-06
B22320-14
CAAAB22320-03EA
108.55
CAD
CAAAB22320-03
CAAAB22320-06
CAAAB22320-14
1-(p-Toluenesulfonyl)indole 95%
1-(p-Toluenesulphonyl)indole
Purity: 95%
CAS No.: 31271-90-6
Molecular Formula: C15H13NO2S
Formula Weight: 271.33
Melting Point: 84-86°
MDL No.: MFCD01165032
Formula:
C₁₅H₁₃NO₂S Melting Pt: 84…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD01165032 CAS Number: 31271-90-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...