N,N'-Diethylhydrazine dihydrochloride tech. 90%, tech.
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22317-06
B22317-14
CAAAB22317-06EA
64.94
CAD
CAAAB22317-06
CAAAB22317-14
N,N'-Diethylhydrazine dihydrochloride tech. 90%, tech.
N,N'-Diethylhydrazine dihydrochloride
Purity: 90%
CAS No.: 7699-31-2
Molecular Formula: CH3CH2NHNHCH2CH3.2HCl
Formula Weight: 159.06
Odor: Odorless
Melting Point: ca 169° dec.
Beilstein Reference: 3668380
MDL No.: MFCD00012523
Formula:
C₄H₁₄Cl₂N₂ Melting Pt: ca 169 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012523 CAS Number: 7699-31-2 EINECS: 206-183-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...