2,2-Dibromomalonamide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22315-14
B22315-22
B22315-09
CAAAAB22315-14EA
183.28
CAD
CAAAAB22315-14
CAAAAB22315-22
CAAAB22315-09
2,2-Dibromomalonamide 98+%
2,2-Dibromomalonamide
Fieser: 1,208
Purity: 98+%
CAS No.: 73003-80-2
Molecular Formula: C3H4Br2N2O2
Formula Weight: 259.89
Melting Point: 202-207°
Beilstein Reference: 1770873
Formula:
C₃H₄Br₂N₂O₂ Melting Pt: 202…207 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014803 CAS Number: 73003-80-2 EINECS: 277-205-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...