2-Hexanoylfuran 99%

Supplier: Thermo Scientific Chemicals

B22306-06 B22306-14
CAAAB22306-06EA 80.9 CAD
CAAAB22306-06 CAAAB22306-14
2-Hexanoylfuran 99%
2-Hexanoylfuran

Purity: 99% CAS No.: 14360-50-0 Molecular Formula: C10H14O2 Formula Weight: 166.22 Flash Point: 107°(225°F) Refractive Index: 1.4890 Odor: Characteristic Boiling Point: 125-127°/15mm Beilstein Reference: 2956 MDL No.: MFCD00037354

Formula: C₁₀H₁₄O₂
Boiling Pt: 125…127 °C/15 mm
Density: 0.994
Flash Pt: 107 °C(225 °F)
Storage Temperature: Ambient
MDL Number: MFCD00037354
CAS Number: 14360-50-0
EINECS: 238-333-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR