Bis(2-mercaptoethyl)sulfide ≥90%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,2'-Thiodiethanethiol, 2-Mercaptoethyl sulfide, 2-Mercaptoethyl Sulphide

B22304-09 B22304-18 B22304-30
CAAAB22304-09EA 149.94 CAD
CAAAB22304-09 CAAAB22304-18 CAAAB22304-30
Bis(2-mercaptoethyl)sulfide ≥90%
Bis(2-mercaptoethyl)sulfide

Purity: 90+% CAS No.: 3570-55-6 Molecular Formula: HSCH2CH2SCH2CH2SH Formula Weight: 154.32 Flash Point: 90°(194°F) Boiling Point: 135-136°/10mm MDL No.: MFCD00004893

Formula: C₄H₁₀S₃
MW: 154.30 g/mol
Boiling Pt: 135…136 °C/10 mm
Density: 1.183
Flash Pt: 90 °C(194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004893
CAS Number: 3570-55-6
EINECS: 222-671-0
UN: 3334

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR