N-Phenylglycine ethyl ester 99%
Supplier: Thermo Scientific Chemicals
B22282-09
B22282-18
CAAAAB22282-09EA
40.45
CAD
CAAAAB22282-09
CAAAAB22282-18
CAAAAB22282-30
N-Phenylglycine ethyl ester 99%
N-Phenylglycine ethyl ester
Purity: 99%
CAS No.: 2216-92-4
Molecular Formula: C6H5NHCH2CO2CH2CH3
Formula Weight: 179.22
Storage Temperature: 2°C to 8°C
Melting Point: 55-58°
MDL No.: MFCD00038315
Formula:
C₁₀H₁₃NO₂ Melting Pt: 55…58 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00038315 CAS Number: 2216-92-4 EINECS: 218-702-2 Merck Index: 14,7292 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...