Diethyl (4-aminobenzyl)phosphonate 99%

Supplier: Thermo Scientific Chemicals

B22279-06 B22279-14 B22279-22
CAAAB22279-06EA 167.48 CAD
CAAAB22279-06 CAAAB22279-14 CAAAB22279-22
Diethyl (4-aminobenzyl)phosphonate 99%
Diethyl (4-aminobenzyl)phosphonate

Purity: 99% CAS No.: 20074-79-7 Molecular Formula: C11H18NO3P Formula Weight: 243.24 Melting Point: 91-94° Beilstein Reference: 2941613

Formula: C₁₁H₁₈NO₃P
MW: 243.24 g/mol
Melting Pt: 91…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00007913
CAS Number: 20074-79-7
EINECS: 243-504-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR