Ranitidine hydrochloride 99%

Supplier: Thermo Scientific Chemicals

B22260-06 B22260-14
CAAAB22260-06EA 110.92 CAD
CAAAB22260-06 CAAAB22260-14
Ranitidine hydrochloride 99%
Ranitidine hydrochloride

An H-2 histamine receptor antagonist


An H-2 histamine receptor antagonist


Applications: An H-2 histamine receptor antagonist

Formula: C₁₃H₂₂N₄O₃S
Melting Pt: ca 134 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00069339
CAS Number: 66357-59-3
EINECS: 266-333-0
Merck Index: 14,8110

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR