BAPTA tetrasodium salt hydrate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: BAPTA-Na4 hydrate, 1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid tetrasodium Salt hydrate, Tetrasodium-1,2-bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetate hydrate

B22258-03 B22258-06 B22258-14
CAAAB22258-03EA 276.5 CAD
CAAAB22258-03 CAAAB22258-06 CAAAB22258-14
BAPTA tetrasodium salt hydrate 98%
BAPTA tetrasodium salt hydrate

BAPTA tetrasodium salt hydrate is a high quality and sensitive compound for calcium signaling studies(investigation of signal transduction and apoptotic cascades, and neuroscience research).


BAPTA, Tetrasodium Salt is useful for spectrophotometric monitoring of extracellular Ca2+ levels, with very little sensitivity to pH. Has also been used for the measurement and control of Ca2+ levels in eggs, chromaffin cells, muscle cells, retinal cells, and pancreatic cells. BAPTA-Na4 is a fluorescent compound utilized to determine free Ca2+ by performing fluorescence emission scans at an excitation wavelength of 299 nm and measuring the emission wavelength at 360 nm.

Formula: C₂₂H₂₀N₂Na₄O₁₀.xH₂O
MW: 564.37 g/mol (anhy)
Storage Temperature: Refrigerator
MDL Number: MFCD02683087
CAS Number: 126824-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR