4-Chloro-6-nitro-o-toluidine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22253-03
B22253-06
CAAAB22253-03EA
31.44
CAD
CAAAB22253-03
CAAAB22253-06
4-Chloro-6-nitro-o-toluidine 98%
4-Chloro-6-nitro-o-toluidine
Purity: 98%
CAS No.: 62790-50-5
Molecular Formula: C7H7ClN2O2
Formula Weight: 186.60
Melting Point: 121-123°
MDL No.: MFCD01320687
Formula:
C₇H₇ClN₂O₂ Melting Pt: 121…123 °C Storage Temperature: Ambient |
MDL Number:
MFCD01320687 CAS Number: 62790-50-5 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...