2-Octen-4-one 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22250-03
B22250-06
CAAAB22250-03EA
131.61
CAD
CAAAB22250-03
CAAAB22250-06
2-Octen-4-one 97%
2-Octen-4-one
Purity: 97%
CAS No.: 4643-27-0
Molecular Formula: C8H14O
Formula Weight: 126.20
Flash Point: 58°(136°F)
Refractive Index: 1.4450
Boiling Point: 65°/15mm
UN No.: 1224
MDL No.: MFCD00061023
Formula:
C₈H₁₄O MW: 126.20 g/mol Boiling Pt: 65 °C/15 mm Density: 0.850 Flash Pt: 58 °C(136 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00061023 CAS Number: 4643-27-0 EINECS: 225-071-7 UN: 1224 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...