1-(2-Chloroethyl)pyrrolidine hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22236-14
B22236-22
CAAAB22236-14EA
81.53
CAD
CAAAB22236-14
CAAAB22236-22
1-(2-Chloroethyl)pyrrolidine hydrochloride 98%
1-(2-Chloroethyl)pyrrolidine hydrochloride
Purity: 98%
CAS No.: 7250-67-1
Molecular Formula: C6H12ClN.HCl
Formula Weight: 170.08
Melting Point: 168-173°
Beilstein Reference: 3677061
MDL No.: MFCD00012718
Formula:
C₆H₁₂ClN Melting Pt: 168…173 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012718 CAS Number: 7250-67-1 EINECS: 230-660-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...