Croconic acid disodium salt 97%

Supplier: Thermo Scientific Chemicals

B22235-01 B22235-03
CAAAB22235-01EA 89.59 CAD
CAAAB22235-01 CAAAB22235-03
Croconic acid disodium salt 97%
Croconic acid disodium salt

Purity: 97% CAS No.: 14379-00-1 Molecular Formula: C5Na2O5 Formula Weight: 186.03 Melting Point: 300° MDL No.: MFCD00191954

Formula: C₅Na₂O₅
MW: 186.03 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00191954
CAS Number: 14379-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR