3-Chloro-2,6-xylidine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Chloro-2,6-dimethylaniline

B22232-09 B22232-18 B22232-30
CAAAB22232-09EA 93.06 CAD
CAAAB22232-09 CAAAB22232-18 CAAAB22232-30
3-Chloro-2,6-xylidine 99%
3-Chloro-2,6-xylidine

Purity: 99% CAS No.: 26829-77-6 Molecular Formula: C8H10ClN Formula Weight: 155.63 Melting Point: 30-32° Boiling Point: 136-138°/17mm MDL No.: MFCD02683086

Formula: C₈H₁₀ClN
MW: 155.63 g/mol
Boiling Pt: 136…138 °C/17 mm
Melting Pt: 30…32 °C
Storage Temperature: Ambient
MDL Number: MFCD02683086
CAS Number: 26829-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR