Sodium nicotinate 98%

Supplier: Thermo Scientific Chemicals
Warning

B22224-22 B22224-36
CAAAB22224-22EA 169.06 CAD
CAAAB22224-22 CAAAB22224-36
Sodium nicotinate 98%
Sodium nicotinate

Purity: 98% CAS No.: 54-86-4 Molecular Formula: C6H4NNaO2 Formula Weight: 145.10 Melting Point: 300°C MDL No.: MFCD00064352

Formula: C₆H₄NNaO₂
MW: 145.10 g/mol
Melting Pt: >300°C
Storage Temperature: Ambient
MDL Number: MFCD00064352
CAS Number: 54-86-4
EINECS: 200-215-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR