4-Ethylcaprylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

B22220-09 B22220-88
CAAAB22220-09EA 199.08 CAD
CAAAB22220-09 CAAAB22220-88
4-Ethylcaprylic acid 97%
4-Ethylcaprylic acid

Purity: 97% CAS No.: 16493-80-4 Molecular Formula: C10H20O2 Formula Weight: 172.27 Flash Point: 100°(212°F) Refractive Index: 1.4390 Boiling Point: 142-143°/10mm MDL No.: MFCD00506494

Formula: C₁₀H₂₀O₂
Boiling Pt: 142…143 °C/10 mm
Density: 0.908
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00506494
CAS Number: 16493-80-4
EINECS: 240-560-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR