1-(3-(Trifluoromethyl)phenoxy)-3-butyn-2-ol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22218-03
B22218-06
CAAAB22218-03EA
92.11
CAD
CAAAB22218-03
CAAAB22218-06
CAAAB22218-14
1-(3-(Trifluoromethyl)phenoxy)-3-butyn-2-ol 98%
1-(3-(Trifluoromethyl)phenoxy)-3-butyn-2-ol
Purity: 98%
CAS No.: 88462-65-1
Molecular Formula: C11H9F3O2
Formula Weight: 230.19
Storage Temperature: -30°C to -10°C
Melting Point: 30-33°
MDL No.: MFCD02683085
Formula:
C₁₁H₉F₃O₂ Melting Pt: 30…33 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02683085 CAS Number: 88462-65-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...