4'-n-Octyloxybiphenyl-4-carboxylic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22216-03
B22216-06
CAAAB22216-03EA
65.57
CAD
CAAAB22216-03
CAAAB22216-06
4'-n-Octyloxybiphenyl-4-carboxylic acid 99%
4'-n-Octyloxybiphenyl-4-carboxylic acid
Purity: 99%
CAS No.: 59748-18-4
Molecular Formula: C21H26O3
Formula Weight: 326.44
MDL No.: MFCD00192369
Formula:
C₂₁H₂₆O₃ MW: 326.44 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00192369 CAS Number: 59748-18-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...