Sodium-1-propanesulfonate 99%

Supplier: Thermo Scientific Chemicals
Warning

B22210-03 B22210-06 B22210-14
CAAAB22210-03EA 62.25 CAD
CAAAB22210-03 CAAAB22210-06 CAAAB22210-14
Sodium-1-propanesulfonate 99%
Sodium-1-propanesulfonate

Purity: 99% CAS No.: 14533-63-2 Molecular Formula: CH3CH2CH2SO3Na Formula Weight: 146.14 MDL No.: MFCD00062546

Formula: C₃H₈O₃S
Storage Temperature: Ambient
MDL Number: MFCD00062546
CAS Number: 14533-63-2
EINECS: 238-564-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR