D-(+)-Melezitose hydrate 99%
Supplier: Thermo Scientific Chemicals
B22209-09
B22209-18
CAAAB22209-09EA
91.64
CAD
CAAAB22209-09
CAAAB22209-18
D-(+)-Melezitose hydrate 99%
D-(+)-Melezitose hydrate
Purity: 99%
CAS No.: 207511-10-2
Molecular Formula: C18H32O16.xH2O
Formula Weight: 504.46 (anhy)
Melting Point: ca 160° dec.
Beilstein Reference: 99539
Optical Rotation: +88°(c=4 in water)
MDL No.: MFCD00149448
Formula:
C₁₈H₃₂O₁₆ Melting Pt: ca 160 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00149448 CAS Number: 207511-10-2 EINECS: 209-894-9 Merck Index: 14,5819 |
Specification Test Results
Optical rotation | +88° (c=4 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...