L-Glutamic acid 5-benzyl ester 99%
Supplier: Thermo Scientific Chemicals
B22184-06
B22184-14
CAAAB22184-03EA
62.39
CAD
CAAAB22184-03
CAAAB22184-06
CAAAB22184-14
L-Glutamic acid 5-benzyl ester 99%
L-Glutamic acid 5-benzyl ester
Purity: 99%
CAS No.: 1676-73-9
Molecular Formula: C12H15NO4
Formula Weight: 237.26
Storage Temperature: 2°C to 8°C
Melting Point: 162-165°
Beilstein Reference: 1885646
Optical Rotation: +23° (c=1 in acetic acid)
MDL No.: MFCD00002633
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...