3-Amino-3-(2-nitrophenyl)propionic acid 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
3-Amino-3-(2-nitrophenyl)propanoic acid
B22176-06
B22176-14
B22176-22
CAAAB22176-06EA
172.22
CAD
CAAAB22176-06
CAAAB22176-14
CAAAB22176-22
3-Amino-3-(2-nitrophenyl)propionic acid 98%
3-Amino-3-(2-nitrophenyl)propionic acid
Purity: 98%
CAS No.: 5678-48-8
Molecular Formula: C9H10N2O4
Formula Weight: 210.19
MDL No.: MFCD00090356
Formula:
C₉H₁₀N₂O₄ MW: 210.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00090356 CAS Number: 5678-48-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...