5,5',6,6'-Tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobiindane 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5,5',6,6'-Tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobisindane

B22170-30 B22170-18
CAAAB22170-30EA 489.8 CAD
CAAAB22170-30 CAAAB22170-18
5,5',6,6'-Tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobiindane 97%
5,5',6,6'-Tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobiindane

Purity: 97% CAS No.: 77-08-7 Molecular Formula: C21H24O4 Formula Weight: 340.42 Melting Point: 300° Beilstein Reference: 2226231 MDL No.: MFCD00021235

Formula: C₂₁H₂₄O₄
MW: 340.42 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00021235
CAS Number: 77-08-7
EINECS: 201-003-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR