Boc-L-tyrosine methyl ester 99%
Supplier: Thermo Scientific Chemicals
B22164-06
B22164-14
B22164-22
CAAAB22164-06EA
107.91
CAD
CAAAB22164-06
CAAAB22164-14
CAAAB22164-22
Boc-L-tyrosine methyl ester 99%
Boc-L-tyrosine methyl ester
Purity: 99%
CAS No.: 4326-36-7
Molecular Formula: C15H21NO5
Formula Weight: 295.34
Melting Point: 105-108°
Optical Rotation: +50° (c=1 chloroform)
MDL No.: MFCD00191181
Formula:
C₁₅H₂₁NO₅ Melting Pt: 105…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191181 CAS Number: 4326-36-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...