4-Cyanophenylhydrazine hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydrazinylbenzonitrile hydrochloride

B22157-06 B22157-14 B22157-22
CAAAB22157-06EA 102.23 CAD
CAAAB22157-06 CAAAB22157-14 CAAAB22157-22
4-Cyanophenylhydrazine hydrochloride 97%
4-Cyanophenylhydrazine hydrochloride

Purity: 97% CAS No.: 2863-98-1 Molecular Formula: C7H7N3.HCl Formula Weight: 169.61 Melting Point: ca 243° dec. Beilstein Reference: 3565486

Formula: C₇H₈ClN₃
MW: 169.61 g/mol
Melting Pt: ∼ 243 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00673994
CAS Number: 2863-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR